Would you like to log in again?
Amersham™ ECL™ Western Blotting Detection Reagent, Cytivia
Supplier: Cytiva
ECL™
RPN2109 RPN2209 RPN2209 RPN2134 RPN2134 RPN2109 RPN2108 RPN2106 RPN2106
95038-566EA 565.93 USD InStock
CA95038-562L CA95038-560L CA95038-566L 95038-566 CA95038-564L 95038-564 95038-562 95038-560 95038-570
Amersham™ ECL™ Western Blotting Detection Reagent, Cytivia
Electrophoresis Reagents Western Blotting Reagents
Enhanced chemiluminescence (ECL) detection reagents for reliable detection of medium and high expression proteins.
  • Ideal for confirmatory Western blotting applications using chemiluminescence detection
  • At least 10-fold more sensitive than colorimetric assays
  • Detectable on Amersham Hyperfilm ECL or with a CCD imager such as ImageQuant LAS 500.
Discover our Western Blotting: tips and troubleshooting eLearning course. Learn approaches to ensure high-quality western blotting courses and receive a certificate upon course completion.
High sensitivity chemiluminescence detection on Western blotting
Amersham ECL Western Blotting Detection Reagents use enhanced luminol-based detection, which is suitable for all routine Western blotting. ECL Western Blotting Detection Reagents, in combination with Amersham Protran membranes, form a complete solution for Western blot applications.
Reliable chemiluminescence detection
In most contemporary ECL systems a luminol peroxide detection reagent is added to the membrane and reacts with the horseradish peroxidase enzyme (HRP) conjugated to the secondary antibody. HRP catalyzes the oxidation of luminol in a multistep reaction and is accompanied by the emission of low-intensity light at 428 nm, which can be measured with light-sensitive X-ray film or with a CCD imager.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty