Would you like to log in again?
Anti-GST Antibody (HRP (Horseradish Peroxidase))
Supplier: Cytiva
RPN1236 RPN1236
95038-580EA 422.88 USD InStock
CA95038-580L 95038-580
Anti-GST Antibody (HRP (Horseradish Peroxidase))
Antibodies
GST is a commonly used expression marker in cloning that is also used for purification and detection of fusion proteins. This highly specific antibody to GST is conjugated to horseradish peroxidase and optimised for use in Western blotting with ECL™ Detection Reagents. The conjugate recognises multiple epitopes of GST and therefore is not reliant on functional GST for fusion protein detection. It has also been cross-absorbed to E. coli proteins so that GST-fusion proteins can be easily and specifically identified against an E. coli background.
  • Offers the advantages of speed, sensitivity, increased safety, and convenience when used with ECL™ detection systems
  • Application tested with ECL™ and ECL™ Plus reagents in Western blotting for sensitive hard copy results
  • Detection of GST is simplified because no secondary antibody is required
  • Binds with high affinity to GST fusion proteins
  • Also suitable for colorimetric detection with diaminobenzidine (DAB) and tetramethylbenzidine (TMB)
GST is a commonly used expression marker in cloning that is also used for purification and detection of fusion proteins. This highly specific antibody to GST is conjugated to horseradish peroxidase and optimized for use in Western blotting with ECL Detection Reagents.
The conjugate recognizes multiple epitopes of GST and therefore is not reliant on functional GST for fusion protein detection. It has also been cross-absorbed to E. coli proteins so that GST-fusion proteins can be easily and specifically identified against an E. coli background.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty