Would you like to log in again?
BBL™ Violet Red Bile Glucose Agar, BD Life Sciences
Supplier: BD Life Sciences
215198 215198
95038-628PK 100.54 USD InStock
CA95038-628 95038-628
BBL™ Violet Red Bile Glucose Agar, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Violet Red Bile Glucose Agar is used for detecting and enumerating Enterobacteriaceae in food and dairy products.
  • Pancreatic digest of gelatin provides carbon, nitrogen, vitamins, and minerals
  • Yeast extract supplies B-complex vitamins
  • Glucose is a carbohydrate
  • Bile salts and crystal violet inhibit gram-positive bacteria
  • Sodium chloride maintains the osmotic balance
  • Agar is the solidifying agent
Violet Red Bile Glucose Agar is recommended for the detection and enumeration of Enterobacteriaceae in food and dairy products. Violet Red Bile Glucose Agar is also listed in the USP as the recommended solid medium for use in the isolation of bile-tolerant gram-negative bacteria from nonsterile pharmaceutical products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty