Would you like to log in again?
Soyabean HiVeg™ Medium, Sterile Powder, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV011G-2.5KG MV011G-500G MV011G-2.5KG MV011G-500G MV011G-5KG MV011G-5KG
89227-186EA 442.68 USD InStock
CA95038-660 CA95038-662 89227-186 95038-662 CA89227-186 95038-660
Soyabean HiVeg™ Medium, Sterile Powder, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Soyabean HiVeg medium, sterile powder is prepared by completely replacing animal-based peptones with vegetable peptones. It is a gamma irradiated sterile powder recommended for the evaluation of sterility in manufacturing process.
The combination of HiVeg hydrolysate and papaic digest of soyabean meal makes this medium nutritious by providing amino acids and long chain peptides for the growth of microorganisms. Dextrose serves as the carbon source and dipotassium phosphate buffers the medium. Sodium chloride maintains the osmotic balance of the medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty