Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-H3 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-778-100
95038-978EA 408 USD InStock
95038-978
Anti-H3 Rabbit Polyclonal Antibody
Antibodies
The nucleosome is made up of four core histone proteins (H2A, H2B, H3 and H4) and is the primary building block of chromatin. The core histones form an octamer, which is made up of one H3-H4 tetramer and two H2A-H2B dimers. The N-terminal tail of core histones undergoes different post-transcriptional modifications including acetylation, phosphorylation, methylation and ubiquitination. These modifications occur in response to cell signal stimuli and have a direct effect on gene expression.

  • For Western Blot

Recognizes human, mouse, rat (weak), bovine (weak) and dog (weak) Histone H3 (dimethyl-Lys9).

Type: Primary
Antigen: HISTONE H3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty