Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Disposable Biopsy Punches, Integra™ Miltex®
Supplier: Integra LifeSciences
33-31-P/25 33-31 33-32-P/25 33-34-P/25 33-31AA 33-31A 33-37 33-36 33-31A-P/25 33-34-P/25 33-31B 33-35 33-34 33-31-P/25 33-34-P/25 33-31AA-P/25 33-38 33-32 33-33
95039-102PK 201.88 USD InStock
CA95039-102 95039-102 95039-104 CA-95039-102 95039-090 21909-136 21909-146 CA-95039-098 95039-092 21909-138 95039-088 21909-140 95039-098 95039-100 21909-144 21909-132 95039-094 21909-142 95039-096
Disposable Biopsy Punches, Integra™ Miltex®
Biopsy Punches
These biopsy punches offer the convenience of single-use disposability without sacrificing quality, feel, or performance
All punches feature seamless, stainless-steel, razor-sharp edges to eliminate torn or pulled biopsies. Ribbed handles provide comfort and control. Punches are color-coded with sizes clearly marked. Sterile, five-year shelf life.
Plunger-system models are available in five sizes and offer the added benefit of effortless ejection of lodged speciments. Non-plunger-system models are available in ten sizes for a wide range of procedures.
Ordering information: Assorted sizes include 10 each of the following diameters: 2 mm, 3 mm, 4 mm, 5 mm, and 6 mm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty