Would you like to log in again?
Sabourauds Chloramphenicol Agar, Modified, Dehydrated Culture Media, Himedia
Supplier: HiMedia Laboratories
M1681-500G M1681-500G
95039-326EA 304.43 USD InStock
CA95039-326 95039-326
Sabourauds Chloramphenicol Agar, Modified, Dehydrated Culture Media, Himedia
Media for Microbiology Dehydrated Media for Microbiology
A selective medium recommended for the isolation of all species of yeast and dermatophytes.
  • Microbiological culture media
  • 500 g poly bottle
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty