Would you like to log in again?
R2A Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1687-500G M1687-500G
95039-342EA 408.37 USD InStock
CA95039-342 95039-342
R2A Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
R2A broth for cultivation and maintenance of heterotrophic bacteria from potable waters.
  • Source of essential growth factors required for metabolism of the bacteria
  • Alternative medium used for the heterotrophic plate counts and for subculturing isolates from potable waters
  • Enables better recovery of anaerobic bacteria from treated waters under different incubation conditions
This medium contains casein acid hydrolysate, yeast extract, biopeptone as a source of essential growth factors required for metabolism of the bacteria.The total bacterial count of drinking water is determined by plate count on a nutritionally rich medium. However all organisms present are not able to grow on them, either because they are slow growers or because they cant grow on that media. For this reason a nutritionally reduced medium was described. R-2A Agar is a modification of this medium. R-2A Agar is an alternative medium used for the heterotrophic plate counts and for subculturing isolates from potable waters. R-2A Agar is also recommended by APHA for pour plate, spread plate and membrane filter technique. R-2A Broth is similar to R-2A Agar. Total count recommended for theb acterial examination of potable waters gives an estimate
of the aerobic and facultatively anaerobic bacteria, which grow best at 35° C in a rich medium. R-2A Broth enables better recovery of these bacteria from treated waters under different incubation conditions. Many bacteria from natural waters, which contain limited nutrients at ambient temperature, grow best on the media with less nutrient levels. They grow better at the temperatures below the routine laboratory incubation temperatures of 35° to 37° C. This medium contains casein acid hydrolysate, yeast extract, biopeptone as source of essential growth factors required for metabolism of the bacteria. Dextrose is the energy source. Starch acts as a neutralizer that neutralizes any toxic metabolites, if present. Phosphate buffers the medium while sodium pyruvate supplies additional nutrition. Magnesium sulphate serves as a source of ions. Due to the presence of the above mentioned ingredients these media allow the growth of stressed and chlorine tolerant bacteria present in treated waters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty