Would you like to log in again?
Middlebrook 7H11 Agar Base without Malachite Green, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M511A-500G M511A-500G
95039-370EA 237.64 USD InStock
95039-370 CA95039-370
Middlebrook 7H11 Agar Base without Malachite Green, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For isolation, cultivation, and determination of antimicrobial susceptibility of mycobacteria.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty