Would you like to log in again?
Dey-Engley Neutralizing Broth, without Bromocresol Purple, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
MU1062-500G MU1062-500G
95039-380EA 306.07 USD InStock
CA95039-380 95039-380
Dey-Engley Neutralizing Broth, without Bromocresol Purple, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For disinfectant testing, where neutralizaton of antiseptics and disinfectants is important for determining bactericidal activity, in accordance with USP.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty