Would you like to log in again?
Letheen Broth Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
MU165-500G MU165-500G
95039-384EA 256.07 USD InStock
CA95039-384 95039-384
Letheen Broth Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the determination of bacterial activity of quaternary ammonium compounds using Escherichia coli or Staphylococcus aureus in accordance with USP.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty