The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based media designed to work in conjunction with or replace standard animal-based protein sources
The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of media can meet the requirements of nearly any microbiology application.