Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gel/PCR DNA Fragments Extraction Kits, IBI Scientific

Supplier: IBI Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

IB47082 IB47020 IB47010 IB47080 IB47030 IB47081
95039-642EA 234.28 USD InStock
95039-642 95039-644 71002-358 71002-356 95039-640 71002-354
Gel/PCR DNA Fragments Extraction Kits, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Kit is designed to recover or concentrate DNA fragments (50bp–10kb) from agarose gel, PCR, or other enzymatic reactions

  • Sample size: Up to 300 mg agarose gel, up to 100 µL PCR products

Note: For research use only.

Caution: Components contain irritant agents. Always wear a lab coat, disposable gloves, and protective goggles during use.

The method uses a chaotropic salt to dissolve agarose gel and denature enzymes. The DNA fragments in the chaotropic salt are bound by the glass fiber matrix of the spin column. The contaminants are washed with a wash buffer (containing ethanol), and the purified DNA fragments are eluted by a low salt elution buffer or TE. Salts, enzymes and unincorporated nucleotides can be effectively removed from the reaction mixture without phenol extraction or alcohol precipitation. Typically, recoveries are up to 90% for gel extraction and up to 95% for PCR clean up. The entire procedure can be completed in 20 minutes and the eluted DNA is ready to use in restriction digestion, ligation, PCR, and sequencing reactions.

Ordering information: User must provide microcentrifuge tubes and absolute ethanol.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty