Would you like to log in again?
Neutralyde™ Formalin Neutralizing Liquid, Azer Scientific
Supplier: Azer Scientific
Neutralyde™
ES767 ES766
95039-814EA 105.26 USD InStock
95039-814 95039-816
Neutralyde™ Formalin Neutralizing Liquid, Azer Scientific
Spill Control Materials
Neutralyde™ Formalin Neutralizing Liquid is designed to safely neutralize and dispose of formalin in the laboratory environment without the threat of exposure to hazardous vapors.
  • No pH adjustment required
  • Phosphate-free
  • Complete neutralization in eight hours
  • One gallon of Neutralyde for five gallons of 10% formalin waste
  • Effective on formaldehyde and glutaraldehyde solutions
  • Safe and gentle reaction
A single gallon of Neutralyde liquid treats up to five gallons of 10% formalin waste and provides a safe and gentle reaction with virtually no risk to personnel.
Neutralyde™ is a phosphate-free, eco-friendly liquid neutralizer that is effective on formaldehyde and glutaraldehyde solutions. Treated waster may be disposed of in sanitary drain.
Ordering information: Odering: Available in 1, 2 1/2 or 5 gallon cubes
Packaging: Packaged in cubitainers equipped with a dispensing spigot.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty