Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA (Deoxyribonucleic acid) (from calf thymus), sonicated

Supplier: Cytiva

27-4563-01 27-4563-01
95040-284EA 681.04 USD InStock
CA95040-284L 95040-284
DNA (Deoxyribonucleic acid) (from calf thymus), sonicated
DNA (Deoxyribonucleic acid)
Genomic DNA sheared by sonication for use in hybridization experiments.

  • Fragmented DNA with a size range from 200 to 6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease

DNA isolated from calf thymus is sonicated to fragment the DNA to an average size of 3000 bp.


Storage Temperature: Freezer
MDL Number: MFCD00130922
CAS Number: 9007-49-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty