Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA (Deoxyribonucleic acid) (from salmon testes), sonicated

Supplier: Cytiva

27-4565-01 27-4565-01
95040-286EA 671.3 USD InStock
95040-286 CA95040-286L
DNA (Deoxyribonucleic acid) (from salmon testes), sonicated
DNA (Deoxyribonucleic acid)
Genomic DNA sheared by sonication for use in hybridization experiments.

  • Fragmented DNA with a size range from 200-6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease
  • Suitable for blocking non-specific binding sites in hybridization experiments to reduce background

Sonicated DNA from salmon testes.


Storage Temperature: Freezer
MDL Number: MFCD00130922
CAS Number: 9007-49-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty