Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA (Deoxyribonucleic acid) (from calf thymus), activated

Supplier: Cytiva

27-4575-01 27-4575-01
95040-288EA 515.3 USD InStock
CA95040-288L 95040-288
DNA (Deoxyribonucleic acid) (from calf thymus), activated
DNA (Deoxyribonucleic acid)
Activated calf thymus DNA at 25 A₂₆₀ units/ml (treated briefly with DNase I) for preparing substrate used in the study of DNA polymerases.

  • Nonmethylated λ DNA (at a concentration of 500 µg/ml) for preparation of molecular weight markers
  • High-quality DNAs from viral and eukaryotic sources used for preparation of assay reagents

Methylated DNA and nonmethylated DNA (at a concentration of 500 µg/ml) for determining methylation sensitivity of restriction enzymes.


MDL Number: MFCD00130922
CAS Number: 9007-49-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty