Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Agarose, highly purified

Supplier: Cytiva

17-0554-02 17-0554-02
95040-294EA 380.18 USD InStock
CA95040-294L 95040-294
Agarose, highly purified
Agarose
Agarose NA has been prepared to give the optimal combination of these properties: Reliable separation of nucleic acids; low inhibition of enzyme activity on recovered material; and high gel strength.

  • Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.
  • Suitable for all analytical separations as well as most preparative methods.

Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications. If separated nucleic acids are to be used as substrates for enzymes (e.g., ligase and restriction endonucleases), a special highly purified agarose such as Agarose NA should be used.
For preparative work, an agarose with low gelling temperature should be used. Agarose Prep has low melting and gelling points.


Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty