Would you like to log in again?
T4 DNA Ligase, Cytiva
Supplier: Cytiva
27-0361-01 27-0361-01
95040-312EA 424.18 USD InStock
CA95040-312L 95040-312
T4 DNA Ligase, Cytiva
Nucleic Acid Reagents Cloning Kits and Ligation
T4 DNA ligase catalyses the formation of a phosphodiester bond between the 5'-phosphoryl group and the 3'-hydroxyl group of two double-stranded DNA fragments. ATP is required for this reaction.
  • For covalently linking DNA fragments in as little as 30 minutes
  • Single dose, ambient-temperature-stable reaction mixes
Catalyzes the formation of a phosphodiester bond between the 5'-phosphoryl group and the 3'-hydroxyl group of two double-stranded DNA fragments. ATP is required for this reaction. Ligation of blunt-ended DNA is greatly stimulated by polyethylene glycol. Applications include self-circularization of linear DNA prior to transformation, ligation of insert to vector, and ligation of synthetic linkers to blunt-ended DNA. Each predispensed, single-reaction, ambient-temperature-stable tube contains a glassified reaction mix which, when reconstituted to a final reaction volume of 20 µl, yields a solution of reaction buffer, T4 DNA ligase, and ATP.
Storage conditions: Room Temperature
Delivery information: Each predispensed, single-reaction, ambient-temperature-stable tube contains a glassified reaction mix which, when reconstituted to a final reaction volume of 20 µl, yields a solution of reaction buffer, T4 DNA ligase, and ATP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty