Would you like to log in again?
Glass Plates, Cytiva
Supplier: Cytiva
80-6190-01 80-6190-01 80-1106-88 80-1106-88
95044-744PK 267.45 USD InStock
CA95044-744 95044-744 CA95040-386 95040-386
Glass Plates, Cytiva
Electrophoresis Systems Vertical Gel Electrophoresis Systems
Polyacrylamide IEF gels can be cast in a cassette system.
The cassette is made up by a glass plate with the film support and an U-frame, and is hold together with clamps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty