Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P53 Mouse Monoclonal Antibody [clone: PAb122]

Supplier: Enzo Life Sciences
KAM-CC002-J KAM-CC002-D KAM-CC002-F
76203-730EA 1720 USD InStock
76203-730 95040-466 95040-464
Anti-P53 Mouse Monoclonal Antibody [clone: PAb122]
Antibodies
p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to Mdm2, SV40 T antigen and human Papilloma virus E6 protein. p53 senses DNA damage and facilitates its repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovary, bladder, colon, lung, and melanoma.

Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone: PAb122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty