Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP60 Mouse Monoclonal Antibody [clone: LK-2]

Supplier: Enzo Life Sciences
ADI-SPA-807-E
95040-498EA 387 USD InStock
95040-498
Anti-HSP60 Mouse Monoclonal Antibody [clone: LK-2]
Antibodies
Hsp60 is a member of the chaperonin family of heat shock proteins, with homologs functioning in the cytosol and mitochondria to fold nascent and aggregated proteins. Hsp60 is the eukaryotic homolog of the E. coli GroEL protein, and forms a multimeric complex in the mitochondria with Hsp10 (Cpn10) to form a large central cavity in which ATP-dependent protein folding takes place. TRiC/CCT, a eukaryotic relative of Hsp60, is expressed in the cytosol and participates in the folding of actin and tubulin substrates, but lacks any association with an Hsp10-like co-factor.

Type: Primary
Antigen: HSP60
Clonality: Monoclonal
Clone: LK-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit, Plant, Pig, Monkey, Hamster, Guinea pig, Dog, Chicken, C. elegans, Bovine, Bacteria

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty