Would you like to log in again?
Bile Salts, Certified, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
CR008-500G CR008-500G
95041-206EA 694.24 USD InStock
CA95041-206 95041-206
Bile Salts, Certified, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
A selectively inhibitory agent in microbiological culture media
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Specially manufactured extract of bile salts is prepared from fresh ox bile.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty