Would you like to log in again?
Casein Enzyme Hydrolysate, Certified, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
CR014-500G CR014-500G
95041-220EA 236.66 USD InStock
CA95041-220 95041-220
Casein Enzyme Hydrolysate, Certified, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
A sterility testing media, diagnostic media, and media for biochemical characterization.
  • Microbiological culture media
  • 500 g poly bottles
Tryptone certified, rich in tryptophan, and used by a wide variety of microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty