Would you like to log in again?
StatSpin® Express 4 Centrifuge, Iris Sample Processing
Supplier: HemoCue
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
StatSpin®
SSH4 SSH4
95041-456EA 7970.19 USD InStock
95041-456 CA95041-456
StatSpin® Express 4 Centrifuge, Iris Sample Processing
Centrifuges Benchtop Centrifuges
This high-speed horizontal centrifuge provides faster turnaround time and greater productivity.
  • Settings for two, three, and five minutes
  • Alarm announces finished cycle
  • Maintenance-free, brushless motor
  • Up to eight samples processed at once
  • Rapid separation of gel tubes
To simplify operations, the StatSpin® Express 4 Centrifuge features three time settings for three, five, and ten minute procedures.
Lid remains locked until the rotor completely stops, and an audible alarm sounds when the cycle is complete. Adapters allow the centrifuge to accommodate tubes from 1.5–10 mL.
Certifications: UL and cUL listed. CE marked.
Ordering information: This centrifuge includes eight-place rotor, eight 7 mL inserts, eight 10 mL inserts, manual latch release tool, hex key tool, and power cord.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty