Would you like to log in again?
HistoZyme
Supplier: Diagnostic BioSystems
K046 K046 K046-50 K046-50
95041-474EA 229.9 USD InStock
CA95041-476 95041-474 CA95041-474 95041-476
HistoZyme
Enzymes
HistoZyme is a proteolytic enzyme solution that results in superior staining over traditionally used enzyme pretreatments, such as pepsin and trypsin.
HistoZyme requires only a brief incubation at room temperature. Performing antigen recovery with HistoZyme, will allow you to dilute the primary antibody 5 to 10 fold further and achieve optimal staining.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty