Would you like to log in again?
PermaRed/AP, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
K049-110 K049 K049-110 K049
95041-484EA 135.32 USD InStock
CA95041-486 CA95041-484 95041-484 95041-486
PermaRed/AP, Diagnostic Biosystems
Assays Cellular Assays
PermaRed/AP is a substrate chromogen system for use with alkaline phosphatase (AP) detection in immunohistochemistry (IHC) or in situ hybridizations (ISH).
PermaRed/AP produces a brilliant dark red color and insoluble in organic solvents; therefore sections can be dehydrated in alcohol, cleared in xylene (or a xylene-substitute), and permanently mounted.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty