Would you like to log in again?
IP ActivFlo Biopsy III Premium Mesh Biopsy Cassettes, Leica
Supplier: Leica
39LC-625-8-L 39LC-625-10-L 39LC-625-4-L 39LC-625-9-L 39VWR-625-5-L 39LC-625-1-L 39LC-625-2-L 39LC-625-11-L 39LC-625-6-L
95041-522CS 402.6 USD InStock
95041-522 95041-524 95041-512 95041-526 95041-514 95041-506 95041-516 95041-508 95041-520
IP ActivFlo Biopsy III Premium Mesh Biopsy Cassettes, Leica
Embedding Cassettes Biopsy Processing Cassettes
IP ActivFlo Biopsy III cassettes feature a patented “Fluidic Flow" pore design for maximum reagent exchange.
  • Suitable for securing the smallest specimens
  • Sloped walls
  • Available in a wide variety of colors
With a 0.26 mm pore size, the cassettes can secure the smallest specimens, eliminating the need for biopsy bags and sponges.
The cassettes also feature sloped walls, creating a basket for small tissue management. These two piece cassettes with hinged lid have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
Ordering information: Cassettes are available with taped packaging for easy printer loading.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty