High performance 25 watt UV tubes for high fluorescence and sensitivity on stained gels
Caution: Transilluminators are powerful sources of ultraviolet radiation which can cause serious damage to unprotected eyes and skin. Read the ultraviolet Caution Information before operating any transilluminators.Caution: Transilluminators are powerful sources of ultraviolet radiation which can cause serious damage to unprotected eyes and skin. Read the ultraviolet Caution Information before operating any transilluminators.
The stainless steel cover frame provides an easy clean surface and reduces cross contamination of samples. Uniform, brilliant back-lit fluorescence illumination is enhanced with Analytik Jena's unique background uniformity screen and UVG long-life filter. Uniformity screen is available on units with mid-range or longwave UV . The transilluminator's powerful cooling fan maintains a low filter surface temperature to prevent sample damage and maintains a constant UV intensity. Select models offer single intensity other models include variable intensity.
Ordering information: The removable and adjustable protective UV blocking cover is included with each transilluminator. Transilluminators also come with a two year warranty.