Would you like to log in again?
Control Slides, Azer Scientific
Supplier: Azer Scientific
ESCS-115 ESCS-118 ESCS-122 ESCS-129 ESCS-109 ESCS-114 ESCS-124 ESCS-125 ESCS-104 ESCS-127 ESCS-130 ESCS-113 ESCS-106 ESCS-116 ESCS-121 ESCS-111 ESCS-117 ESCS-107 ESCS-126
95042-310PK 108.65 USD InStock
95042-310 95042-332 95042-288 95042-286 95042-296 95042-282 95042-292 95042-318 95042-328 95042-306 95042-316 95042-304 95042-326 95042-302 95042-324 95042-334 95042-322 95042-300 95042-308
Control Slides, Azer Scientific
Slides Microscopy Control Slides
Control slides are designed for use in monitoring stain performance in the Pathology lab.
  • Permanent positive charge for improved readability
  • Control slides with paraffin-embedded human tissue sections
  • Ten unstained Unifrost slides
  • One stained slide
All slides feature a permanent positive charge that securely anchors all types of sections and improves the quality of preparations and readability
Includes 10 unstained Unifrost microscope slides and one additional slide, which is stained with the proper procedure to demonstrate and validate the expected result.
Azer Scientific's control slides are carefully prepared with paraffin-embedded human tissue sections.
Packaging: 11 per pack
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty