Would you like to log in again?
UVP Transilluminator, Analytik Jena
Supplier: Analytik Jena
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
95-0458-02 95-0456-01 95-0459-02 95-0452-01 95-0413-02 95-0449-02 95-0447-02 95-0459-01 95-0459-01 95-0458-01 95-0417-01 95-0457-01 95-0447-01 95-0414-01 95-0414-02 95-0447-01 95-0459-02 95-0455-01 95-0458-01 95-0449-02 95-0457-02 95-0449-01 95-0456-01 95-0417-01 95-0413-01 95-0449-01 95-0455-01 95-0456-02 95-0413-01 95-0452-02 95-0452-01 95-0414-01 95-0417-02 95-0457-01
95042-376EA 3760.7 USD InStock
95042-376 CA95042-386 95042-396 CA95042-388 95042-374 CA95042-366 95042-372 95042-394 95042-370 95042-392 95042-358 CA95042-382 CA95042-384 CA95042-362 95042-356 95042-378 95042-390 95042-386 95042-364 95042-384 CA95042-356 CA95042-378 95042-362 95042-360 CA95042-358 95042-382 95042-380 CA95042-390 CA95042-370 CA95042-394 95042-368 95042-366 95042-388 CA95042-374
UVP Transilluminator, Analytik Jena
Transilluminators UV Transilluminators
UVP offers a wide range of filter sizes and single or multiple UV wavelength transilluminators in a compact, benchtop size.
  • Increased ultraviolet output and intensity
  • Fast lamp start up with no light flicker
  • Universal power supply with reduced electrical consumption
  • 8-Watt tubes provide longer tube life
  • Lighter weight
  • Improved gel visualization and gel documentation
  • Clear hinged and adjustable UV blocking cover included
With the 365 nm longwave UV, view gels for extended periods of time, avoiding photonicking damage. The 302nm mid-range UV increases the fluorescence for photo documentation and the 254 nm shortwave UV will irradiate samples.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty