Would you like to log in again?
VWR® Autoclavable PP Bags
Supplier: VWR
VWR®
1102-0810 1102-2436 1102-1224 1102-0815 1102-1012 1102-0812 1102-1010 1102-2430 1102-1015
95042-552PK 119.65 USD InStock
95042-552 95042-563 95042-550 95042-548 95042-558 95042-568 95042-556 95042-554 95042-564
VWR® Autoclavable PP Bags
Bags Waste Disposal Bags
Transparent 2 mil (0.05 mm) thick polypropylene bags are suitable for autoclaving hazardous and nonhazardous materials and waste.
  • Can withstand autoclaving temperatures up to 135 °C (275 °F)
Twist ties (included) allow proper closure for bag expansion and venting during sterilization.
Certifications: Meet strength requirements in ASTM D 1709-04 for 165g dart drop test.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty