Would you like to log in again?
KOD Xtreme™ Hot Start DNA Polymerase, MilliporeSigma
Supplier: MilliporeSigma
Xtreme™
71975-3 71975-3
95042-700EA 854.04 USD InStock
CA95042-700 95042-700
KOD Xtreme™ Hot Start DNA Polymerase, MilliporeSigma
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The KOD Xtreme™ Hot Start DNA Polymerase is a ready-to-use reaction or routine PCR amplification, and optimized for difficult targets
Efficiently amplifies up to 90% GC-content DNA templates, genomic targets up to 24 kbp and phage/Lambda targets up to 40 kbp.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty