Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKG Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-PK005-F
95042-926EA 499 USD InStock
95042-926
Anti-PKG Rabbit Polyclonal Antibody
Antibodies
Cyclic GMP-dependent protein kinases (PKGs or cGKs) are classified into two types, PKGI and PKGII. Studies have shown that PKGs are highly homologous to PKAs; phosphorylation of cellular proteins by both families of kinases leads to alterations in calcium mobilization, protein phosphatase activity, ion channel function, gene transcription, smooth muscle contractility, and platelet aggregation.

Type: Primary
Antigen: PKG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty