Would you like to log in again?
HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
3089803IH25-40 SH30898.02 SH30898.01 SH30898.02HI SH30898.02 SH30898.03 3089803IH25-40 SH30898.03 SH30898.01 SH30898.03IR25-40 SH30898.03IR25-40 SH30898.03HI SH30898.02HI SH30898.03HI
95042-942EA 308.98 USD InStock
95042-942 CA95042-942 95042-944 CA76534-640 76534-640 76324-914 CA76324-914 CA76324-912 CA76635-870 76635-870 76324-912 CA95042-944 95042-948 CA95042-948
HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
Sera (Serum)
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS).
  • Processed via protein G chromatography
  • Enhance cell culture growth
  • U.S. sourced
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS), processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty