Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
UVP DigiDoc-It® Imaging System, Analytik Jena
Supplier: Analytik Jena
DigiDoc-It®
97-0246-01 97-0243-02 97-0105-01 97-0244-01 97-0244-02 97-0246-02 97-0243-01
95043-000EA 8609.01 USD InStock
95043-000 95042-998 95043-006 14220-784 95043-008 95043-002 95043-004
UVP DigiDoc-It® Imaging System, Analytik Jena
Photodocumentation Systems
The DigiDoc-It® system is ideal for image acquisition and documentation of gels, plates and membranes. System includes a digital color camera, hood enclosure, UV diopter and ethidium bromide filter, and image capture software.
The digital camera features 5.1 megapixels, 640x480 to 2592x1944 resolution, 4x optical zoom lens, UV filters, and a diopter
The optional Doc-It® 1D analysis software loads on a separate computer for analysis of gels, plates, and membranes. Features include automatic lane and band detection, quantitative analysis of lanes and bands, multiple MW calibration, background correction, image enhancement, and reporting capabilities.
Imaging of nonfluorescent gels requires a UV to white converter plate. The Visi-Blue™ convertor plate converts ultraviolet to 480nm blue light for use with SYBR® Green, SYPRO® Orange, and EGFP stains. Both plates have an area of 21x26 cm (8 1/4x10 1/4").
The image capture software loads on a user’s computer configured with Windows® 98, ME, or 2000 operating systems with a USB interface. The software controls camera functions including live image preview, focus and exposure settings, and image saturation warnings. It also features enhancement and annotation, detailed and configurable reporting, and history tracking.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty