Would you like to log in again?
Cocoon Environmental Enrichment Bedding, Ketchum
Supplier: Ketchum Manufacturing
XKA 2600 087
95043-336PK 971.25 USD InStock
95043-336
Cocoon Environmental Enrichment Bedding, Ketchum
Animal Supplies Animal Environmental Enrichment
Designed to benefit the health and contentment of rodents used for research, the Cocoon offers animals the oppurtunity to engage in social activity and fulfill nesting instincts
The small, lightweight, cylindrical form of the Cocoon makes it easily moveable, increasing the interaction between the animal, the enrichment product, and the environment. The cylinder is constructed of short cotton fibers that are safe for the animal and its offspring. Once torn, the Cocoon can be modified to create a vaporous cloud which can be used to create a nest. Cocoon is whitened without chlorine. Packaged 6,000 cylinders per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty