Would you like to log in again?
Non-Ionic Detergent Solutions, G-Biosciences
Supplier: G-Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DG019 DG017 786-513 786-521 786-515 DG021 786-522 786-519 786-351 DG021 DG025 786-512 786-517 DG020 786-517 DG020 786-352 786-521 786-514 786-512 DG022 786-511 DG019 786-516 786-351 DG017 786-352 786-518 786-511 786-514 DG015 DG023 DG022 786-516 786-515 DG018 786-520 786-522 DG015 DG024 DG016 786-520 DG016 DG026 786-519 DG025 786-513 DG023 DG024 DG026 786-518 DG018
95057-562EA 115.08 USD InStock
95057-562 95057-560 CA95043-472 CA95043-470 CA82022-994 CA95043-478 CA82022-992 CA95043-476 CA95043-474 CA82022-998 CA82022-996 CA82023-000 CA82023-002 95043-492 95043-490 82022-996 82022-994 82022-992 82022-998 82023-002 CA82023-012 82023-004 CA82023-014 82023-000 CA82023-010 95043-484 CA82023-008 95043-482 CA95043-492 CA82023-004 95043-480 CA95043-490 CA82023-006 95043-488 95043-486 CA95057-560 82023-014 82023-010 82023-012 CA95057-562 CA95043-482 95043-474 CA95043-480 95043-472 82023-006 95043-470 82023-008 CA95043-488 CA95043-486 95043-478 CA95043-484 95043-476
Non-Ionic Detergent Solutions, G-Biosciences
Cleaning Agents Detergents and Surface Cleansers
Non-ionic detergents have a hydrophilic head group that is uncharged and are preferred for their ability to break lipid-lipid and lipid-protein interactions
The non-ionic deteregents are supplied as a general Research Grade, Proteomic Grade (PG) Solutions and 2D-Detergents™. The Proteomic Grade (PG) Solutions have ultra low aldehyde (<100μM) and peroxide (<50μM) concentrations to reduce the effects of peroxidase and carbonyl compounds that negatively interact with membrane proteins. The 2D-Detergents™ have low conductivity (<10μS) and ultra low aldehyde (<100μM) and peroxide (<50μM) concentrations.
G-Biosciences Non-Ionic Detergent Solutions include: Triton® X-100, Triton® X-114, Brij® 35, Brij® 58, Tween® 20, Tween® 80, Nonidet® P-40 Substitute, Octyl ß Glucoside, MEGA 8, MEGA 9, MEGA 10, BigCHAP, and Deoxy Big CHAP.
They have limited ability to break protein-protein interactions and are often referred to as non-denaturing detergents and are used to isolate biologically active membrane proteins.
Ordering information: Accessories not included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty