Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRP55 Mouse Monoclonal Antibody [clone: FMC 75]

Supplier: Enzo Life Sciences
SPA-601-488-F
95043-742EA 639 USD InStock
95043-742
Anti-CRP55 Mouse Monoclonal Antibody [clone: FMC 75]
Antibodies
The multifunctional, multi -compartmental protein Calreticulin (Crt) functions as a soluble molecular chaperone of new or misfolded proteins, as well as a Ca2+-binding protein. Most abundant in the ER lumen, Crt expression also occurs in other membrane-bound organelles, the cell surface, and extracellularly. Also known as CRP-55, calregulin and HACBP (high affinity calcium-binding protein), Crt contains the ER-retrieval sequence, KDEL, and is the solub le paralog of the ER membrane protein Calnexin (Cnx). Crt's three domains include a 180 residue N-terminal domain, a proline-rich Pdomainresidues 189 -288) that binds Ca2+ with high affinity and shares homology with Cnx and calmegin, and a 110 residue C-terminal domain that binds Ca2+ with low affinity but high capacity. The P-domain may interact with the co-chaperone ERp57 (Grp58), a thiol reductase. The NMR structure of the P -domain consists of an extended hairpin that appears to form a curved protrusion from the Crt core domain. Both Crt and its membrane bound homolog CNX interact with proteins and glycoproteins possessing monoglucosylated N -glycans. The Crt/Cnx cycle promotes correct folding, inhibits aggregation of folding intermediates, blocks premature oligomerization, regulates ER degradation, and prevents incompletely folded glycoproteins from exiting to the Golgi complex. Crt also appears to function as an auto-antigen in systemic lupus erythematosus, rheumatoid arthritis, celiac disease, complete congenital heart block, and halothane hepatitis. A diversity of additional functions attributed to Crt includes adhesion, blood function, and cardiac and neuronal development gene expression.

Type: Primary
Antigen: CRP55
Clonality: Monoclonal
Clone: FMC 75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty