Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MT Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-550-D ADI-SPA-550-F
95043-876EA 556 USD InStock
95043-876 95043-874
Anti-MT Mouse Monoclonal Antibody
Antibodies
Metallothioneins (MTs) are a class of ubiquitous, low molecular weight, cysteine-rich, metal binding proteins that function in the metabolism of zinc and copper. MT protects against exposure to toxic metals such as mercury and cadmium as well as a variety of other stress conditions. There are four mammalian MT isoforms: MT-I, -II, -III, -IV, each of which is encoded by individual genes. MT expression is induced by heavy metal cations, starvation, elevated temperature, lipopolysaccharide, inflammatory cytokines, free radicals, and ionizing radiation.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse, rat, fish, horse and rabbit metallothionein. Detects a band of ~15kDa by Western blot.

Type: Primary
Antigen: MT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty