Would you like to log in again?
Neutralizing Reagent, (27 ml), Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-80-1476
95043-880EA 153 USD InStock
95043-880 CA95043-880
Neutralizing Reagent, (27 ml), Enzo Life Sciences
Buffers Biological Buffers
Neutralizing buffer for low pH solutions, for use with cyclic nucleotide and isoprostane kits.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty