Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Pyruvate dehydrogenase Kinase 1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-PK112-F
95043-894EA 499 USD InStock
95043-894
Anti-Pyruvate dehydrogenase Kinase 1 Rabbit Polyclonal Antibody
Antibodies
PDK-1 (Pyruvate dehydrogenase kinase-1) is a 48 kDa protein kinase. It is a member of the mitrochondrial matrix protein kinase family and is related to the histidine protein kinases found in prokaryotes. PDK-1 is responsible for phosphorylation and concomitant inactivation of pyruvate dehydrogenase.

Type: Primary
Antigen: Pyruvate dehydrogenase kinase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit, Pig, Monkey, Dog

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty