Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STAT3 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-TF022-C KAP-TF022F
95043-918EA 201 USD InStock
95043-918 CA95045-996
Anti-STAT3 Rabbit Polyclonal Antibody
Antibodies
Stats (Signal transducers and activators of transcription) are a family of cytoplasmic latent transcription factors that are activated to regulate gene expression in response to a large number of extracellular signaling polypeptides including cytokines, interferons, and growth factors. After phosphorylation by JAK tyrosine kinases, Stats enter the nucleus to regulate transcription of many different genes.

  • For Western Blot

Recognizes human, mouse (weak), rat (weak), bovine (weak) and pig (weak) Stat3.

Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Pig, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty