Would you like to log in again?
Stop Solution, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-80-0247 ADI-80-0248 80-0247
10049-208EA 163.09 USD InStock
10049-208 CA95043-924 95043-924
Stop Solution, Enzo Life Sciences
Assays ELISAs
Assay stop solution for AP EIA/ELISA
  • Liquid in form
This is a solution of TRIS-sodium phosphate in water.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty