Would you like to log in again?
Midi Fast-Ion Plasmid Kits, Endotoxin Free, IBI Scientific
Supplier: IBI Scientific
IB47113 IB47111 IB47112
95039-658EA 368.97 USD InStock
95039-658 95043-944 95043-942
Midi Fast-Ion Plasmid Kits, Endotoxin Free, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
IBI's Midi Fast-Ion Plasmid Kits are endotoxin free and use pre-packed ion-exchange resin columns to purify plasmid or cosmid DNA from bacterial cultures
  • Sample Size: 50 ml high-copy plasmid/100 ml low-copy plasmid
  • Operation: Gravity Flow Column
  • Expectant Yield: 200 µg of plasmid DNA
  • Operation Time: 120 minutes or less
The Midi Fast Ion Plasmid Kit uses pre-packed ion-exchange resin columns to purify plasmid or cosmid DNA from bacterial cultures. In this process, the modified alkaline lysis method and RNase treatment are used for creating cleared lysate with minimal genomic DNA and RNA contaminants. Using a gravity-flow procedure, the plasmid DNA in crude lysate has been bound to the column. The contaminants can be washed off with a wash buffer. Finally, the purified DNA is eluted by a high salt buffer and then precipitated with isopropanol for desalting. The entire procedure can be completed in less than two hours. The purified plasmid DNA is suitable for transfection, sequencing reactions, PCR, and in-vitro transcription.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty