Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Transferrin Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
VAP-EN004-F VAP-EN004-D VAP-EN004D VAP-EN004F
95045-168EA 499 USD InStock
95045-168 CA95045-166 95045-166 CA95045-168
Anti-Transferrin Rabbit Polyclonal Antibody
Antibodies
Transferrin is a serum glycoprotein involved in iron transport. Transferrin exists in two forms, the iron-free Apotransferrin, which binds two Fe3+ ions to generate Ferrotransferrin. Apotransferrin remains tightly bound to its receptor until the complex is recycled to the cell surface, where Apotransferrin is released into the extracellular space to recruit more Fe3+ ions.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, dog and guinea pig Transferrin. Detects a band of ~76kDa by Western blot.

Type: Primary
Antigen: TRANSFERRIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty