Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Src Kinase Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-678-100 905-678-100
95045-822EA 421 USD InStock
CA95045-822 95045-822
Anti-Src Kinase Mouse Monoclonal Antibody
Antibodies
Src is a member of the tyrosine kinase family that was originally identified as the oncogene of Rous sarcoma virus. The catalytic domain contains a tyrosine autophosphorylation site that increases kinase activity when phosphorylated. The Src family of tyrosine kinase is involved in a variety of receptor-induced signal transduction pathways such as immune recognition rec

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat Src. Detects a band of ~60kDa by Western blot.

Type: Primary
Antigen: SRC KINASE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty