Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LSD1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-769-100 905-769-100
95045-850EA 421 USD InStock
95045-850 CA95045-850
Anti-LSD1 Mouse Monoclonal Antibody
Antibodies
Lysine-specific demethylase 1 (LSD1, AOF2, BHC110) is a nuclear amine oxidase homolog, and was the first such protein to display definitive activity in epigenetic modulation as a histone demethylase. LSD1 demethylates histone H3 at mono- or di-methylated lysine residues. Depending on associated co-factors, LSD1-mediated demethylation can lead to transcriptional repression or activation.

  • For Western Blot

Recognizes human, mouse and rat LSD1/AOF2.

Type: Primary
Antigen: LSD1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty