Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SEPT5 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-781-100 905-781-100
95045-874EA 361 USD InStock
CA95045-874 95045-874
Anti-SEPT5 Mouse Monoclonal Antibody
Antibodies
Septin 5 (Sept5/CDCrel-1) is a member of the Septin family of cytoskeletal GTPases. Septin 5 expression is primarily associated with synaptic vesicles in cells of the nervous system where it acts to inhibit exocytosis, although it has also been characterized as an inhibitor of aggregation and release of storage granules in platelets. Phosphorylation of Septin 5 at Ser17 by Cdk5-p35 has been shown to mediate the interaction of Septin 5 with the SNARE protein Syntaxin-1A, regulating neurotransmitter release. Targeted disruption of the Septin 5 gene in mice failed to produce an obvious neurological phenotype, indicating its loss may be overcome by functional redundancy of other septins. Septin 5 is one of several Parkin (an E3 ubiquitin ligase) associated substrates whose accumulation results in cellular toxicity related to the onset of autosomal recessive Parkinson Disease.

  • For Western Blot

Recognizes human, mouse and rat Septin 5/Cdcrel-1. Detects a band of ~43kDa by Western blot.

Type: Primary
Antigen: SEPT5
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty