Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACT1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
CSA-400F ADI-CSA-400-F CSA-400D ADI-CSA-400-D
95045-898EA 499 USD InStock
CA95045-898 95045-898 CA95045-896 95045-896
Anti-ACT1 Rabbit Polyclonal Antibody
Antibodies
Actin is one of the major proteins (~42 kDa) of the cytoskeleton. It regulates contractile potential in muscle tissues and helps to control the shape as well as the motility of non-muscle cells. Actin is expressed in all eukaryotic cells which makes it a useful loading control in Western blotting experiments. In higher eukaryotes, there are six isoforms of actin: Three alpha-actins (α-skeletal, α-cardiac, α-smooth muscle), one β-actin (β-non-muscle) and two γ-actins (γ-smooth muscle and γ-non-muscle). Actin isoforms show >90% overall sequence homology.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Actin. Detects a band of ~42kDa by Western blot.

Type: Primary
Antigen: ACT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty